2,5-Bis(allyloxy)terephthalohydrazide 98%

Supplier: Ambeed
Warning

A1156874-100MG A1156874-1G A1156874-250MG A1156874-5G
76675-210EA 23 USD
76675-210 76675-212 76675-214 76675-216
2,5-Bis(allyloxy)terephthalohydrazide 98%
2,5-Bis(allyloxy)terephthalohydrazide

Precaución: for Research Use Only

Formula: C₁₄H₁₈N₄O₄
MDL Number: MFCD34165586
CAS Number: 2227151-69-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR