4,4',4'',4'''-(4,4'-(1,4-Phenylene)bis(pyridine-6,4,2-triyl))tetrabenzoic acid 98%

Supplier: Ambeed
Warning

A1156872-100MG A1156872-1G A1156872-250MG
76675-204EA 33 USD
76675-204 76675-206 76675-208
4,4',4'',4'''-(4,4'-(1,4-Phenylene)bis(pyridine-6,4,2-triyl))tetrabenzoic acid 98%
4,4',4'',4'''-(4,4'-(1,4-Phenylene)bis(pyridine-6,4,2-triyl))tetrabenzoic acid

Precaución: for Research Use Only

Formula: C₄₄H₂₈N₂O₈
MDL Number: MFCD34165585
CAS Number: 1836122-41-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR