1,4-Bis((1H-1,2,4-triazol-1-yl)methyl)benzene 98%

Supplier: Ambeed
Warning

A1156864-250MG A1156864-25G A1156864-5G
76675-198EA 28.65 USD
76675-198 76675-200 76675-202
1,4-Bis((1H-1,2,4-triazol-1-yl)methyl)benzene 98%
1,4-Bis((1H-1,2,4-triazol-1-yl)methyl)benzene

Precaución: for Research Use Only

Formula: C₁₂H₁₂N₆
MW: 240.2639 g/mol
CAS Number: 143131-66-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR