1,3,5-Tris(1H-benzo[d]imidazol-2-yl)benzene 98%

Supplier: Ambeed
Warning

A1156862-100MG A1156862-1G A1156862-250MG A1156862-5G
76675-190EA 19.1 USD
76675-190 76675-192 76675-194 76675-196
1,3,5-Tris(1H-benzo[d]imidazol-2-yl)benzene 98%
1,3,5-Tris(1H-benzo[d]imidazol-2-yl)benzene

Precaución: for Research Use Only

Formula: C₂₇H₁₈N₆
MW: 426.4720 g/mol
MDL Number: MFCD02031506
CAS Number: 351437-96-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR