[1,1':3',1''-Terphenyl]-4,4''-dicarboxylic acid, 5'-(4-carboxyphenyl)-2'-hydroxy- 98%

Supplier: Ambeed
Warning

A1156853-100MG A1156853-250MG
76675-178EA 29 USD
76675-178 76675-180
[1,1':3',1''-Terphenyl]-4,4''-dicarboxylic acid, 5'-(4-carboxyphenyl)-2'-hydroxy- 98%
[1,1':3',1''-Terphenyl]-4,4''-dicarboxylic acid, 5'-(4-carboxyphenyl)-2'-hydroxy-

Precaución: for Research Use Only

Formula: C₂₇H₁₈O₇
CAS Number: 1353118-75-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR