1,4-Di(1H-1,2,4-triazol-1-yl)benzene 98%

Supplier: Ambeed
Warning

A1156849-1G A1156849-250MG A1156849-5G
76675-172EA 92.3 USD
76675-172 76675-174 76675-176
1,4-Di(1H-1,2,4-triazol-1-yl)benzene 98%
1,4-Di(1H-1,2,4-triazol-1-yl)benzene

Precaución: for Research Use Only

Formula: C₁₀H₈N₆
MW: 212.2107 g/mol
CAS Number: 1435710-71-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR