3'',4'',5'',6''-Tetrakis(4'-carboxy[1,1'-biphenyl]-4-yl)[1,1':4',1'':2'',1''':4''',1''''-quinquephenyl]-4,4''''-dicarboxylic acid 98%
Supplier: Ambeed
|
Warning
|
A1156843-100MG
A1156843-1G
A1156843-250MG
76675-166EA
87
USD
76675-166
76675-168
76675-170
3'',4'',5'',6''-Tetrakis(4'-carboxy[1,1'-biphenyl]-4-yl)[1,1':4',1'':2'',1''':4''',1''''-quinquephenyl]-4,4''''-dicarboxylic acid 98%
3'',4'',5'',6''-Tetrakis(4'-carboxy[1,1'-biphenyl]-4-yl)[1,1':4',1'':2'',1''':4''',1''''-quinquephenyl]-4,4''''-dicarboxylic acid
Precaución: for Research Use Only
Formula:
C₈₄H₅₄O₁₂ MW: 1255.3200 g/mol |
MDL Number:
MFCD31700796 CAS Number: 1374404-53-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...