Tris(4-(4H-1,2,4-triazol-4-yl)phenyl)amine 98%

Supplier: Ambeed
Warning

A1156841-250MG A1156841-5G A1156841-1G
76675-160EA 41.38 USD
76675-160 76675-162 77620-276
Tris(4-(4H-1,2,4-triazol-4-yl)phenyl)amine 98%
Tris(4-(4H-1,2,4-triazol-4-yl)phenyl)amine

Precaución: for Research Use Only

Formula: C₂₄H₁₈N₁₀
MW: 446.4667 g/mol
CAS Number: 2001066-82-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR