3-(4,6-Diphenyl-1,3,5-triazin-2-yl)-9-phenyl-9H-carbazole 98%

Supplier: Ambeed
Warning

A1156822-100MG A1156822-1G A1156822-250MG
76675-128EA 57.29 USD
76675-128 76675-130 76675-132
3-(4,6-Diphenyl-1,3,5-triazin-2-yl)-9-phenyl-9H-carbazole 98%
3-(4,6-Diphenyl-1,3,5-triazin-2-yl)-9-phenyl-9H-carbazole

Precaución: for Research Use Only

Formula: C₃₃H₂₂N₄
MW: 474.5546 g/mol
MDL Number: MFCD31657478
CAS Number: 1313391-57-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR