2,4,6-Tris(4-(bromomethyl)phenyl)-1,3,5-triazine 98%
Supplier: Ambeed
|
Danger
|
A1156818-10G
A1156818-1G
A1156818-25G
A1156818-5G
76675-120EA
582
USD
76675-120
76675-122
76675-124
76675-126
2,4,6-Tris(4-(bromomethyl)phenyl)-1,3,5-triazine 98%
2,4,6-Tris(4-(bromomethyl)phenyl)-1,3,5-triazine
Precaución: for Research Use Only
Formula:
C₂₄H₁₈Br₃N₃ |
MDL Number:
MFCD27965501 CAS Number: 1169964-41-3 UN: 3261 ADR: 8,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...