[1,1':3',1''-Terphenyl]-4,4''-dicarboxylic acid, 2'-amino-5'-(4-carboxyphenyl)- 98%

Supplier: Ambeed
Warning

A1156797-1G A1156797-250MG
76675-106EA 108 USD
76675-106 77866-096
[1,1':3',1''-Terphenyl]-4,4''-dicarboxylic acid, 2'-amino-5'-(4-carboxyphenyl)- 98%
[1,1':3',1''-Terphenyl]-4,4''-dicarboxylic acid, 2'-amino-5'-(4-carboxyphenyl)-

Precaución: for Research Use Only

Formula: C₂₇H₁₉NO₆
MDL Number: MFCD34165563
CAS Number: 1414662-67-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR