2,4,6-Tris(3-(pyridin-3-yl)phenyl)-1,3,5-triazine 98%

Supplier: Ambeed
Warning

A1156750-100MG A1156750-1G A1156750-250MG
76675-088EA 41 USD
76675-088 76675-090 76675-092
2,4,6-Tris(3-(pyridin-3-yl)phenyl)-1,3,5-triazine 98%
2,4,6-Tris(3-(pyridin-3-yl)phenyl)-1,3,5-triazine

Precaución: for Research Use Only

Formula: C₃₆H₂₄N₆
CAS Number: 939430-26-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR