1,3-Bis(4-(tert-butyl)phenyl)propan-2-one 98%

Supplier: Ambeed
Warning

A1156722-100MG A1156722-1G A1156722-250MG A1156722-5G
76675-076EA 21 USD
76675-076 76675-078 76675-080 76675-082
1,3-Bis(4-(tert-butyl)phenyl)propan-2-one 98%
1,3-Bis(4-(tert-butyl)phenyl)propan-2-one

Precaución: for Research Use Only

Formula: C₂₃H₃₀O
CAS Number: 64321-35-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR