Ir(dFFppy)2(dtbbpy)PF6 98%

Supplier: Ambeed
Warning

A1156572-100MG A1156572-1G A1156572-250MG
76675-058EA 102.91 USD
76675-058 76675-060 76675-062
Ir(dFFppy)2(dtbbpy)PF6 98%
Ir(dFFppy)2(dtbbpy)PF6

Precaución: for Research Use Only

Formula: C₄₀H₃₄F₆IrN₄·F₆P
MW: 1021.9000 g/mol
MDL Number: MFCD31807347
CAS Number: 2042201-18-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR