3,6-Dimethoxythieno[3,2-b]thiophene 98%

Supplier: Ambeed
Warning

A1156560-1G A1156560-250MG
76675-048EA 160 USD
76675-048 76675-050
3,6-Dimethoxythieno[3,2-b]thiophene 98%
3,6-Dimethoxythieno[3,2-b]thiophene

Precaución: for Research Use Only

Formula: C₈H₈O₂S₂
MDL Number: MFCD20529143
CAS Number: 850233-79-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR