Methyl 2-(2-((((tert-butoxycarbonyl)amino)oxy)methyl)phenyl)-2-(methoxyimino)acetate 95%

Supplier: Ambeed
Danger

A1156511-1G A1156511-250MG
76675-042EA 316.15 USD
76675-042 76675-044
Methyl 2-(2-((((tert-butoxycarbonyl)amino)oxy)methyl)phenyl)-2-(methoxyimino)acetate 95%
Methyl 2-(2-((((tert-butoxycarbonyl)amino)oxy)methyl)phenyl)-2-(methoxyimino)acetate

Precaución: for Research Use Only

Formula: C₁₆H₂₂N₂O₆
MW: 338.3557 g/mol
CAS Number: 2459977-70-7
UN: 1325
ADR: 4.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR