s-Indacene-1,3,5,7(2H,6H)-tetraone 99%

Supplier: Ambeed
Warning

A1156378-100MG A1156378-1G
76675-030EA 58.35 USD
76675-030 77570-946
s-Indacene-1,3,5,7(2H,6H)-tetraone 99%
s-Indacene-1,3,5,7(2H,6H)-tetraone

Precaución: for Research Use Only

Formula: C₁₂H₆O₄
MW: 214.17 g/mol
MDL Number: MFCD01956452
CAS Number: 53910-13-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR