2-Bromo-4-iododibenzo[b,d]furan 99%

Supplier: Ambeed
Warning

A1156364-100MG A1156364-5G
76675-028EA 19.1 USD
76675-028 77620-254
2-Bromo-4-iododibenzo[b,d]furan 99%
2-Bromo-4-iododibenzo[b,d]furan

Precaución: for Research Use Only

Formula: C₁₂H₆BrIO
MW: 372.98 g/mol
MDL Number: MFCD32669933
CAS Number: 1401068-25-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR