4-Hexyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene-2-carbaldehyde 98%

Supplier: Ambeed
Warning

A1156355-1G A1156355-250MG
76675-020EA 425.43 USD
76675-020 76675-022
4-Hexyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene-2-carbaldehyde 98%
4-Hexyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene-2-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₇H₂₇BO₃S
MW: 322.2705 g/mol
MDL Number: MFCD32695464
CAS Number: 1402132-40-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR