2,5-Bis(trimethylstannyl)thiophene 99%

Supplier: Ambeed
Danger

A1156338-1G A1156338-250MG
76675-018EA 108 USD
76675-018 77866-077
2,5-Bis(trimethylstannyl)thiophene 99%
2,5-Bis(trimethylstannyl)thiophene

Precaución: for Research Use Only

Formula: C₁₀H₂₀SSn₂
MDL Number: MFCD01940352
CAS Number: 86134-26-1
UN: 3146
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR