2,2'-((2Z,2'Z)-((4,4,9,9-Tetrahexyl-4,9-dihydro-s-indaceno[1,2-b:5,6-b']dithiophene-2,7-diyl)bis(methanylylidene))bis(3-oxo-2,3-dihydro-1H-indene-2,1-diylidene))dimalononitrile 99%

Supplier: Ambeed
Warning

A1156310-100MG A1156310-1G A1156310-250MG A1156310-5G
76675-008EA 81 USD
76675-008 76675-010 76675-012 77620-244
2,2'-((2Z,2'Z)-((4,4,9,9-Tetrahexyl-4,9-dihydro-s-indaceno[1,2-b:5,6-b']dithiophene-2,7-diyl)bis(methanylylidene))bis(3-oxo-2,3-dihydro-1H-indene-2,1-diylidene))dimalononitrile 99%
2,2'-((2Z,2'Z)-((4,4,9,9-Tetrahexyl-4,9-dihydro-s-indaceno[1,2-b:5,6-b']dithiophene-2,7-diyl)bis(methanylylidene))bis(3-oxo-2,3-dihydro-1H-indene-2,1-diylidene))dimalononitrile

Precaución: for Research Use Only

Formula: C₆₆H₆₆N₄O₂S₂
CAS Number: 1883441-92-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR