3,6-Bis(5-bromothiophen-2-yl)pyrrolo[3,4-c]pyrrole-1,4(2H,5H)-dione 98%

Supplier: Ambeed
Warning

A1156269-100MG A1156269-250MG A1156269-1G
76675-000EA 48 USD
76675-000 76675-002 77566-080
3,6-Bis(5-bromothiophen-2-yl)pyrrolo[3,4-c]pyrrole-1,4(2H,5H)-dione 98%
3,6-Bis(5-bromothiophen-2-yl)pyrrolo[3,4-c]pyrrole-1,4(2H,5H)-dione

Precaución: For research use only.

Formula: C₁₄H₆Br₂N₂O₂S₂
CAS Number: 777079-55-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR