19-(3-Iodopropyl)heptatriacontane ≥97%

Supplier: Ambeed
Warning

A1156240-100MG A1156240-250MG
76674-996EA 53.05 USD
76674-996 76674-998
19-(3-Iodopropyl)heptatriacontane ≥97%
19-(3-Iodopropyl)heptatriacontane

Precaución: for Research Use Only

Formula: C₄₀H₈₁I
MW: 688.9756 g/mol
MDL Number: MFCD32012260
CAS Number: 1413918-74-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR