Ethyl 5-fluoro-7-nitro-1H-indole-2-carboxylate 97%

Supplier: Ambeed
Warning

A1156125-100MG A1156125-1G A1156125-250MG
76674-974EA 78.51 USD
76674-974 76674-976 76674-978
Ethyl 5-fluoro-7-nitro-1H-indole-2-carboxylate 97%
Ethyl 5-fluoro-7-nitro-1H-indole-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₉FN₂O₄
MW: 252.1986 g/mol
MDL Number: MFCD20923215
CAS Number: 913287-12-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR