3-Amino-4-(4-methylphenyl)pyrrolidine, 3-BOC protected hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl N-[4-(4-methylphenyl)pyrrolidin-3-yl]carbamate hydrochloride, tert-Butyl (4-(p-tolyl)pyrrolidin-3-yl)carbamate hydrochloride

A1156057-1G A1156057-250MG
76674-970EA 368.14 USD
76674-970 76674-972
3-Amino-4-(4-methylphenyl)pyrrolidine, 3-BOC protected hydrochloride 97%
3-Amino-4-(4-methylphenyl)pyrrolidine, 3-BOC protected hydrochloride

Precaución: for Research Use Only

Formula: C₁₆H₂₄N₂O₂·ClH
MW: 312.835 g/mol
MDL Number: MFCD32111679
CAS Number: 2367002-73-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR