2-Phenoxy-1-phenylpropane-1,3-diol 97%

Supplier: Ambeed
Warning

A1155911-1G A1155911-250MG
76674-956EA 271.6 USD
76674-956 76674-958
2-Phenoxy-1-phenylpropane-1,3-diol 97%
2-Phenoxy-1-phenylpropane-1,3-diol

Precaución: for Research Use Only

Formula: C₁₅H₁₆O₃
MW: 244.2857 g/mol
MDL Number: MFCD32067774
CAS Number: 70110-65-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR