N-(2-Chloro-4-pyridinyl)benzamide 98%

Supplier: Ambeed
Warning

A1155759-1MG A1155759-25MG A1155759-50MG
76674-950EA 27.59 USD
76674-950 76674-952 76674-954
N-(2-Chloro-4-pyridinyl)benzamide 98%
N-(2-Chloro-4-pyridinyl)benzamide

Precaución: for Research Use Only

Formula: C₁₂H₉ClN₂O
MW: 232.67 g/mol
MDL Number: MFCD23131104
CAS Number: 591754-12-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR