3-Amino-5-(trifluoromethoxy)phenol 97%

Supplier: Ambeed
Warning

A1155675-100MG A1155675-1G A1155675-250MG A1155675-5G
76674-942EA 78.51 USD
76674-942 76674-944 76674-946 76674-948
3-Amino-5-(trifluoromethoxy)phenol 97%
3-Amino-5-(trifluoromethoxy)phenol

Precaución: for Research Use Only

Formula: C₇H₆F₃NO₂
MW: 193.1233 g/mol
MDL Number: MFCD18390132
CAS Number: 1261650-51-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR