Dimethyl 2-chloropyridine-3,4-dicarboxylate 97%

Supplier: Ambeed
Warning

A1155661-100MG A1155661-1G A1155661-250MG
76674-936EA 118.83 USD
76674-936 76674-938 76674-940
Dimethyl 2-chloropyridine-3,4-dicarboxylate 97%
Dimethyl 2-chloropyridine-3,4-dicarboxylate

Precaución: for Research Use Only

Formula: C₉H₈ClNO₄
MW: 229.6171 g/mol
MDL Number: MFCD28128962
CAS Number: 521980-84-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR