6-Amino-2,4-dichloronicotinonitrile 97%

Supplier: Ambeed
Warning

A1155639-1G A1155639-250MG A1155639-5G
76674-930EA 266.29 USD
76674-930 76674-932 76674-934
6-Amino-2,4-dichloronicotinonitrile 97%
6-Amino-2,4-dichloronicotinonitrile

Precaución: for Research Use Only

Formula: C₆H₃Cl₂N₃
MW: 188.0141 g/mol
MDL Number: MFCD28966659
CAS Number: 1589541-56-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR