Tris(1,10-phenanthroline)ruthenium(II) bis(hexafluorophosphate) 98%

Supplier: Ambeed
Warning

A1155401-100MG A1155401-1G A1155401-250MG A1155401-50MG A1155401-5G
76674-916EA 23 USD
76674-916 76674-918 76674-920 76674-922 77620-232
Tris(1,10-phenanthroline)ruthenium(II) bis(hexafluorophosphate) 98%
Tris(1,10-phenanthroline)ruthenium(II) bis(hexafluorophosphate)
Formula: C₃₆H₂₄F₁₂N₆P₂Ru
MDL Number: MFCD28369263
CAS Number: 60804-75-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR