4-Ethylphenyl hydrogen sulfate 97%

Supplier: Ambeed
Danger

Synonyms: 4-Ethylphenyl sulphate

A1155177-100MG A1155177-250MG A1155177-1G
76674-904EA 161 USD
76674-904 77620-212 77866-055
4-Ethylphenyl hydrogen sulfate 97%
4-Ethylphenyl hydrogen sulfate

Precaución: for Research Use Only

Formula: C₈H₁₀O₄S
MW: 202.2276 g/mol
CAS Number: 85734-98-1
UN: 3261
ADR: 8 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR