7-(Difluoromethoxy)-N-(trans-4-(2-hydroxypropan-2-yl)cyclohexyl)quinoline-3-carboxamide 99%

Supplier: Ambeed
Warning

A1155033-10MG A1155033-1MG A1155033-25MG A1155033-50MG A1155033-5MG
76674-894EA 29 USD
76674-894 76674-896 76674-898 76674-900 76674-902
7-(Difluoromethoxy)-N-(trans-4-(2-hydroxypropan-2-yl)cyclohexyl)quinoline-3-carboxamide 99%
7-(Difluoromethoxy)-N-(trans-4-(2-hydroxypropan-2-yl)cyclohexyl)quinoline-3-carboxamide

Precaución: for Research Use Only

Formula: C₂₀H₂₄F₂N₂O₃
MDL Number: MFCD32067944
CAS Number: 2101626-26-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR