9-(1-Cyclopropylethyl)-N2-(2-(dimethylamino)ethyl)-N6-(3-(trifluoromethyl)benzyl)-9H-purine-2,6-diamine 99%
Supplier: Ambeed
|
Warning
|
A1155032-10MG
A1155032-1MG
A1155032-25MG
A1155032-50MG
76674-886EA
39
USD
76674-886
76674-888
76674-890
76674-892
9-(1-Cyclopropylethyl)-N2-(2-(dimethylamino)ethyl)-N6-(3-(trifluoromethyl)benzyl)-9H-purine-2,6-diamine 99%
9-(1-Cyclopropylethyl)-N2-(2-(dimethylamino)ethyl)-N6-(3-(trifluoromethyl)benzyl)-9H-purine-2,6-diamine
Precaución: for Research Use Only
Formula:
C₂₂H₂₈F₃N₇ |
MDL Number:
MFCD32067989 CAS Number: 2342583-66-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...