3-(5-(2-(1H-Imidazol-1-yl)acetyl)-2-isopropoxyphenyl)-2-((4-(2-(4-chlorophenoxy)acetyl)piperazin-1-yl)methyl)quinazolin-4(3H)-one 98%

Supplier: Ambeed
Warning

A1155014-10MG A1155014-1MG A1155014-5MG A1155014-50MG A1155014-100MG
76674-838EA 206 USD
76674-838 76674-840 76674-842 77620-210 77866-052
3-(5-(2-(1H-Imidazol-1-yl)acetyl)-2-isopropoxyphenyl)-2-((4-(2-(4-chlorophenoxy)acetyl)piperazin-1-yl)methyl)quinazolin-4(3H)-one 98%
3-(5-(2-(1H-Imidazol-1-yl)acetyl)-2-isopropoxyphenyl)-2-((4-(2-(4-chlorophenoxy)acetyl)piperazin-1-yl)methyl)quinazolin-4(3H)-one

Precaución: for Research Use Only

Formula: C₃₅H₃₅ClN₆O₅
MDL Number: MFCD31813799
CAS Number: 1801530-11-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR