(S)-6-(4-Chlorophenyl)-2-(3-fluorophenyl)-N-(1-hydroxypropan-2-yl)-3-oxo-2,3-dihydropyridazine-4-carboxamide 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
BAY-218
A1155004-10MG
A1155004-1MG
A1155004-25MG
A1155004-5MG
76674-830EA
101.85
USD
76674-830
76674-832
76674-834
76674-836
(S)-6-(4-Chlorophenyl)-2-(3-fluorophenyl)-N-(1-hydroxypropan-2-yl)-3-oxo-2,3-dihydropyridazine-4-carboxamide 98%
(S)-6-(4-Chlorophenyl)-2-(3-fluorophenyl)-N-(1-hydroxypropan-2-yl)-3-oxo-2,3-dihydropyridazine-4-carboxamide
Precaución: for Research Use Only
Formula:
C₂₀H₁₇ClFN₃O₃ MW: 401.82 g/mol |
MDL Number:
MFCD31736303 CAS Number: 2162982-11-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...