(1R)-4-((S)-4-Acryloyl-2-methylpiperazin-1-yl)-6-fluoro-7-(2-fluoro-6-hydroxyphenyl)-1-(2-isopropyl-4-methylpyridin-3-yl)pyrido[2,3-d]pyrimidin-2(1H)-one 98%

Supplier: Ambeed
Warning

Synonyms: Sotorasib

A1155000-1G A1155000-250MG A1155000-100MG A1155000-50MG A1155000-5G A1155000-25MG A1155000-5MG
76674-826EA 269.47 USD
76674-826 76674-828 77566-068 77599-660 77595-914 77620-206 77620-208
(1R)-4-((S)-4-Acryloyl-2-methylpiperazin-1-yl)-6-fluoro-7-(2-fluoro-6-hydroxyphenyl)-1-(2-isopropyl-4-methylpyridin-3-yl)pyrido[2,3-d]pyrimidin-2(1H)-one 98%
(1R)-4-((S)-4-Acryloyl-2-methylpiperazin-1-yl)-6-fluoro-7-(2-fluoro-6-hydroxyphenyl)-1-(2-isopropyl-4-methylpyridin-3-yl)pyrido[2,3-d]pyrimidin-2(1H)-one

Precaución: For research use only.

Formula: C₃₀H₃₀F₂N₆O₃
MW: 560.59 g/mol
MDL Number: MFCD32062763
CAS Number: 2296729-00-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR