Dimesityl(trifluoromethyl)sulfonium trifluoromethanesulfonate 95%

Supplier: Ambeed
Warning

A1154976-5G A1154976-1G A1154976-250MG
76674-824EA 421.18 USD
76674-824 77549-280 77592-260
Dimesityl(trifluoromethyl)sulfonium trifluoromethanesulfonate 95%
Dimesityl(trifluoromethyl)sulfonium trifluoromethanesulfonate

Precaución: for Research Use Only

Formula: C₂₀H₂₂F₆O₃S₂
MW: 488.5073 g/mol
MDL Number: MFCD31619197
CAS Number: 1895006-01-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR