1,3-Bis(4-bromophenyl)propan-2-one 97%

Supplier: Ambeed
Warning

A1154848-1G A1154848-5G
76674-804EA 24 USD
76674-804 77570-924
1,3-Bis(4-bromophenyl)propan-2-one 97%
1,3-Bis(4-bromophenyl)propan-2-one

Precaución: for Research Use Only

Formula: C₁₅H₁₂Br₂O
MDL Number: MFCD12031109
CAS Number: 54523-47-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR