N-([1,1'-Biphenyl]-4-yl)-N-(4'-bromo-[1,1'-biphenyl]-4-yl)-9,9-dimethyl-9H-fluoren-2-amine 97%

Supplier: Ambeed
Warning

A1154826-1G A1154826-5G A1154826-25G
76674-800EA 5 USD
76674-800 76674-802 77866-047
N-([1,1'-Biphenyl]-4-yl)-N-(4'-bromo-[1,1'-biphenyl]-4-yl)-9,9-dimethyl-9H-fluoren-2-amine 97%
N-([1,1'-Biphenyl]-4-yl)-N-(4'-bromo-[1,1'-biphenyl]-4-yl)-9,9-dimethyl-9H-fluoren-2-amine

Precaución: for Research Use Only

Formula: C₃₉H₃₀BrN
MDL Number: MFCD28975089
CAS Number: 1268621-99-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR