1-(3-Methylbut-2-en-1-yl)guanidine hydrochloride 97%

Supplier: Ambeed
Warning

A1154820-10MG A1154820-1MG A1154820-25MG A1154820-50MG
76674-792EA 73.21 USD
76674-792 76674-794 76674-796 76674-798
1-(3-Methylbut-2-en-1-yl)guanidine hydrochloride 97%
1-(3-Methylbut-2-en-1-yl)guanidine hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₄ClN₃
MW: 163.65 g/mol
MDL Number: MFCD32069582
CAS Number: 2368870-39-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR