1,4-Bis(2-(pyridin-4-yl)vinyl)benzene 98%

Supplier: Ambeed
Warning

A1154807-1G A1154807-250MG A1154807-5G
76674-786EA 177.18 USD
76674-786 76674-788 76674-790
1,4-Bis(2-(pyridin-4-yl)vinyl)benzene 98%
1,4-Bis(2-(pyridin-4-yl)vinyl)benzene

Precaución: for Research Use Only

Formula: C₂₀H₁₆N₂
MW: 284.3544 g/mol
CAS Number: 3095-81-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR