1,2,4,5-Tetrakis(4-carboxyphenyl)benzene 98%

Supplier: Ambeed
Warning

A1154801-100MG A1154801-10G A1154801-1G A1154801-250MG A1154801-5G
76674-762EA 14 USD
76674-762 76674-764 76674-766 76674-768 76674-770
1,2,4,5-Tetrakis(4-carboxyphenyl)benzene 98%
1,2,4,5-Tetrakis(4-carboxyphenyl)benzene

Precaución: for Research Use Only

Formula: C₃₄H₂₂O₈
MDL Number: MFCD16621436
CAS Number: 1078153-58-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR