4,4-(1,10-Phenanthroline-2,9-diyl)bis[benzaldehyde] 98%

Supplier: Ambeed
Warning

A1154798-100MG A1154798-250MG A1154798-1G A1154798-5G
76674-758EA 36 USD
76674-758 76674-760 77570-922 77866-044
4,4-(1,10-Phenanthroline-2,9-diyl)bis[benzaldehyde] 98%
4,4-(1,10-Phenanthroline-2,9-diyl)bis[benzaldehyde]

Precaución: for Research Use Only

Formula: C₂₆H₁₆N₂O₂
MDL Number: MFCD31539985
CAS Number: 120085-99-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR