4',4''',4''''',4'''''''-Methanetetrayltetrakis(([1,1'-biphenyl]-4-carboxylic acid)) 98%

Supplier: Ambeed
Warning

A1154797-100MG A1154797-1G A1154797-250MG
76674-752EA 42.44 USD
76674-752 76674-754 76674-756
4',4''',4''''',4'''''''-Methanetetrayltetrakis(([1,1'-biphenyl]-4-carboxylic acid)) 98%
4',4''',4''''',4'''''''-Methanetetrayltetrakis(([1,1'-biphenyl]-4-carboxylic acid))

Precaución: for Research Use Only

Formula: C₅₃H₃₆O₈
MW: 800.8482 g/mol
CAS Number: 1208241-38-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR