4',4''',4'''''-Nitrilotris(([1,1'-biphenyl]-4-carboxylic acid)) 98%

Supplier: Ambeed
Warning

Synonyms: Antibacterial agent 18

A1154795-1G A1154795-250MG A1154795-5G
76674-744EA 64.72 USD
76674-744 76674-746 77620-192
4',4''',4'''''-Nitrilotris(([1,1'-biphenyl]-4-carboxylic acid)) 98%
4',4''',4'''''-Nitrilotris(([1,1'-biphenyl]-4-carboxylic acid))

Precaución: for Research Use Only

Formula: C₃₉H₂₇NO₆
MW: 605.6348 g/mol
MDL Number: MFCD31700772
CAS Number: 1239602-35-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR