[3,3-Bipyridine]-6,6-dicarboxaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: [3,3'-Bipyridine]-6,6'-dicarbaldehyde

A1154793-100MG A1154793-250MG A1154793-1G A1154793-5G
76674-742EA 41.38 USD
76674-742 77592-258 77595-912 77620-190
[3,3-Bipyridine]-6,6-dicarboxaldehyde 98%
[3,3-Bipyridine]-6,6-dicarboxaldehyde

Precaución: for Research Use Only

Formula: C₁₂H₈N₂O₂
MW: 212.2041 g/mol
MDL Number: MFCD28338178
CAS Number: 1264748-06-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR