Triphenylene-2,3,6,7,10,11-hexaamine hexahydrochloride 97%

Supplier: Ambeed
Warning

A1154789-100MG A1154789-1G A1154789-250MG A1154789-5G
76674-722EA 100.79 USD
76674-722 76674-724 76674-726 76674-728
Triphenylene-2,3,6,7,10,11-hexaamine hexahydrochloride 97%
Triphenylene-2,3,6,7,10,11-hexaamine hexahydrochloride

Precaución: for Research Use Only

Formula: C₁₈H₂₄Cl₆N₆
MW: 537.1414 g/mol
MDL Number: MFCD31700744
CAS Number: 1350518-27-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR