Tris(4-(pyridin-4-yl)phenyl)amine 98%

Supplier: Ambeed
Warning

A1154786-1G A1154786-250MG A1154786-5G
76674-716EA 289.63 USD
76674-716 76674-718 76674-720
Tris(4-(pyridin-4-yl)phenyl)amine 98%
Tris(4-(pyridin-4-yl)phenyl)amine

Precaución: for Research Use Only

Formula: C₃₃H₂₄N₄
MW: 476.5705 g/mol
MDL Number: MFCD31700774
CAS Number: 1366291-62-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR