1,3,6,8-Tetra(pyridin-4-yl)pyrene 98%

Supplier: Ambeed
Warning

A1154783-100MG A1154783-250MG
76674-708EA 56 USD
76674-708 76674-710
1,3,6,8-Tetra(pyridin-4-yl)pyrene 98%
1,3,6,8-Tetra(pyridin-4-yl)pyrene

Precaución: for Research Use Only

Formula: C₃₆H₂₂N₄
MW: 510.5867 g/mol
MDL Number: MFCD31700776
CAS Number: 1402429-80-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR